missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole, 98%

$21.59 - $86.68

Chemical Identifiers

CAS 50257-40-4
Molecular Formula C18H26N2O2S
Molecular Weight (g/mol) 334.478
MDL Number MFCD00005283
InChI Key AGGRGODMKWLSDE-UHFFFAOYSA-N
Synonym 1-2,4,6-triisopropylphenyl sulfonyl-1h-imidazole, 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1-2,4,6-triisopropylphenylsulfonyl imidazole, tpsi, 1-2',4',6'-triisopropylbenzenesulphonyl imidazole, 1-2,4,6-tri propan-2-yl phenyl sulfonylimidazole, 1-2,4,6-triisopropylphenyl sulfonyl-1h-imidazole, tpsi 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1h-imidazole, 1-2,4,6-tris 1-methylethyl phenyl sulfonyl, 1-2,4,6-trisopropylbenzenesulfonyl imidazole
PubChem CID 521274
IUPAC Name 1-[2,4,6-tri(propan-2-yl)phenyl]sulfonylimidazole
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N2C=CN=C2)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6048203 View Documents Alfa Aesar™
H6048203
1g Each for $21.59
 
AAH6048206 View Documents Alfa Aesar™
H6048206
5g Each for $86.68
 
Specifications

Chemical Identifiers

50257-40-4
334.478
AGGRGODMKWLSDE-UHFFFAOYSA-N
521274
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N2C=CN=C2)C(C)C
C18H26N2O2S
MFCD00005283
1-2,4,6-triisopropylphenyl sulfonyl-1h-imidazole, 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1-2,4,6-triisopropylphenylsulfonyl imidazole, tpsi, 1-2',4',6'-triisopropylbenzenesulphonyl imidazole, 1-2,4,6-tri propan-2-yl phenyl sulfonylimidazole, 1-2,4,6-triisopropylphenyl sulfonyl-1h-imidazole, tpsi 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1h-imidazole, 1-2,4,6-tris 1-methylethyl phenyl sulfonyl, 1-2,4,6-trisopropylbenzenesulfonyl imidazole
1-[2,4,6-tri(propan-2-yl)phenyl]sulfonylimidazole

Specifications

118°C to 122°C
50257-40-4
MFCD00005283
AGGRGODMKWLSDE-UHFFFAOYSA-N
1-[2,4,6-tri(propan-2-yl)phenyl]sulfonylimidazole
521274
98%
1g
C18H26N2O2S
1-2,4,6-triisopropylphenyl sulfonyl-1h-imidazole, 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1-2,4,6-triisopropylphenylsulfonyl imidazole, tpsi, 1-2',4',6'-triisopropylbenzenesulphonyl imidazole, 1-2,4,6-tri propan-2-yl phenyl sulfonylimidazole, 1-2,4,6-triisopropylphenyl sulfonyl-1h-imidazole, tpsi 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1h-imidazole, 1-2,4,6-tris 1-methylethyl phenyl sulfonyl, 1-2,4,6-trisopropylbenzenesulfonyl imidazole
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N2C=CN=C2)C(C)C
334.478
334.48
1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C