missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 138370050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1,2,4,5-Tetramethylbenzene | |
196.0°C to 197.0°C | |
Authentic | |
Glass bottle | |
C6H2(CH3)4 | |
05,431 | |
durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren | |
SQNZJJAZBFDUTD-UHFFFAOYSA-N | |
1,2,4,5-tetramethylbenzene | |
7269 | |
134.22 |
95-93-2 | |
73°C | |
97% min. (GC) | |
C10H14 | |
5g | |
15,3515 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene | |
CC1=CC(=C(C=C1C)C)C | |
134.22 | |
CHEBI:38978 | |
97+% |
Chemical Identifiers
95-93-2 | |
134.22 | |
durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren | |
CHEBI:38978 | |
CC1=CC(=C(C=C1C)C)C |
C10H14 | |
SQNZJJAZBFDUTD-UHFFFAOYSA-N | |
7269 | |
1,2,4,5-tetramethylbenzene |
Safety and Handling
GHS H Statement
Flammable solid.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
GHS Signal Word: Danger
EINECSNumber : 202-465-7
RUO â Research Use Only