Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,4,5-Tetrakis(isopropylthio)benzene, 97%

$145.70 - $528.88

Chemical Identifiers

Chemical Name or Material 1,2,4,5-Tetrakis(isopropylthio)benzene
CAS 74542-69-1
Molecular Formula C18H30S4
Formula Weight 374.68
MDL Number MFCD04039784
Synonym 1,2,4,5-Tetra(isopropylthio)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2526103

  SDS
alfa aesar™
B2526103
1g Each for $145.70

AAB2526106

  SDS
alfa aesar™
B2526106
5g Each for $528.88
Description & Specifications

Specifications

Formula Weight 374.68
Chemical Name or Material 1,2,4,5-Tetrakis(isopropylthio)benzene
Assay 97%
Molecular Formula C18H30S4
MDL Number MFCD04039784
CAS 74542-69-1
Synonym 1,2,4,5-Tetra(isopropylthio)benzene
Melting Point 76° to 78°C
Quantity 1g