Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,4,5-Tetrachlorobenzene, 98%

Chemical Identifiers

Chemical Name or Material 1,2,4,5-Tetrachlorobenzene
CAS 95-94-3
Molecular Formula C6H2Cl4
Formula Weight 215.89
MDL Number MFCD00000549
UN Number UN3077
Beilstein 1618315
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1431809

 
alfa aesar™
A1431809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1431818

 
alfa aesar™
A1431818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1431830

 
alfa aesar™
A1431830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.89
UN Number UN3077
TSCA TSCA
CAS 95-94-3
Boiling Point 245°C
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 1 Warning
MDL Number MFCD00000549
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Flash Point 155°C (311°F)
Quantity 10g
Odor Unpleasant
Beilstein 1618315
Molecular Formula C6H2Cl4
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Chemical Name or Material 1,2,4,5-Tetrachlorobenzene
Melting Point 138° to 142°C
RTECS Number DB9450000

Applications
It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.

Solubility
Insoluble in water. Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.