Learn More
1,2,4,5-Tetrabromobenzene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 352401000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2,4,5-Tetrabromobenzene | |
636-28-2 | |
100.0 | |
Beige to Brown | |
96% min. (GC) | |
C6H2Br4 | |
MFCD00000063 | |
benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo | |
QCKHVNQHBOGZER-UHFFFAOYSA-N | |
1,2,4,5-tetrabromobenzene | |
12486 | |
97% |
97% | |
96.0 | |
177.0°C to 182.0°C | |
Authentic | |
Glass bottle | |
C6H2Br4 | |
100 g | |
Solubility in water: insoluble. | |
BrC1=CC(Br)=C(Br)C=C1Br | |
393.70 | |
393.7 | |
Crystalline Powder, Crystals or Needles |
Chemical Identifiers
636-28-2 | |
393.70 | |
QCKHVNQHBOGZER-UHFFFAOYSA-N | |
1,2,4,5-tetrabromobenzene |
C6H2Br4 | |
MFCD00000063 | |
12486 | |
BrC1=CC(Br)=C(Br)C=C1Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause long lasting harmful effects to aquatic life.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 211-253-3
TSCA : TSCA
RUO – Research Use Only