Learn More
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139285000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 2, 4-1H-Triazole | |
99.4 | |
119°C to 122°C | |
260°C | |
Authentic | |
Plastic bottle | |
MFCD00005228 | |
26, 13 | |
15, 9766 | |
Solubility in water: 1250g/L (20°C). Other solubilities: soluble in chloroform,xylene,ethylacetate and,alcohol | |
C1=NC=NN1 | |
69.06 | |
CHEBI:35550 | |
99.50% |
288-88-0 | |
100.0 | |
White | |
140°C | |
99.4% min. (HPLC) | |
C2H3N3 | |
500 g | |
01,1188; 02,423 | |
1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van | |
NSPMIYGKQJPBQR-UHFFFAOYSA-N | |
1H-1,2,4-triazole | |
9257 | |
69.06 | |
Crystalline Powder or Flakes |
Chemical Identifiers
288-88-0 | |
69.06 | |
NSPMIYGKQJPBQR-UHFFFAOYSA-N | |
9257 | |
1H-1,2,4-triazole |
C2H3N3 | |
MFCD00005228 | |
1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van | |
CHEBI:35550 | |
C1=NC=NN1 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Suspected of damaging the unborn child.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr
GHS Signal Word: Warning
EINECSNumber : 206-022-9
RUO – Research Use Only