Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3-Tris(2-cyanoethoxy)propane, 97%

Chemical Identifiers

Chemical Name or Material 1,2,3-Tris(2-cyanoethoxy)propane
CAS 2465-93-2
Formula Weight 251.29
MDL Number MFCD00019868
UN Number UN3276
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3670214

 
alfa aesar™
3670214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3670222

 
alfa aesar™
3670222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.29
Chemical Name or Material 1,2,3-Tris(2-cyanoethoxy)propane
TSCA TSCA
Physical Form Liquid
Assay 97%
MDL Number MFCD00019868
CAS 2465-93-2
UN Number UN3276
Health Hazard 1 Danger
Density 1.107g/mL
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.