missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3-Trimethyl-3H-benzo[e]indole, TCI America™

Manufacturer:  TCI America T311925G

 View more versions of this product

Catalog No. T311925G



Specifications

Specifications

1,2,3-Trimethyl-3H-benzo[e]indole
25g
C15H15N
1,2,3-Trimethylnaphtho[1,2-d]pyrrole
CC1=C(N(C2=C1C3=CC=CC=C3C=C2)C)C
209.292
209.29
≥98.0% (GC)
Undesignated
881219-73-4
MFCD06651515
KQMROBBTYFXFOV-UHFFFAOYSA-N
1,2,3-trimethylbenzo[e]indole
16439627
Crystal-Powder at 20°C

Chemical Identifiers

881219-73-4
209.292
KQMROBBTYFXFOV-UHFFFAOYSA-N
16439627
CC1=C(N(C2=C1C3=CC=CC=C3C=C2)C)C
C15H15N
MFCD06651515
1,2,3-Trimethylnaphtho[1,2-d]pyrrole
1,2,3-trimethylbenzo[e]indole
Safety and Handling

Safety and Handling

TSCA : No

Documents