missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3-Trimethoxybenzene, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 160211000

 View more versions of this product

Catalog No. AC160211000



Specifications

Specifications

1, 2, 3-Trimethoxybenzene
112°C
Glass bottle
0.5% max.
241.0°C
42.0°C to 45.0°C
98%
97.5
97.5% min. (GC)
C6H3(OCH3)3
06, 1081
Solubility in water: insoluble
COC1=C(C(=CC=C1)OC)OC
168.19
CHEBI:86529
Powder
1.1120g/mL
Authentic
1.112
1% max.
Beige to White
100g
634-36-6
100.0
C9H12O3
MFCD00008358
1,2,3-trimethoxy benzene
CRUILBNAQILVHZ-UHFFFAOYSA-N
1,2,3-trimethoxybenzene
12462
168.19
98%

Chemical Identifiers

634-36-6
168.19
CRUILBNAQILVHZ-UHFFFAOYSA-N
12462
1,2,3-trimethoxybenzene
C9H12O3
MFCD00008358
1,2,3-trimethoxy benzene
CHEBI:86529
COC1=C(C(=CC=C1)OC)OC
Safety and Handling

Safety and Handling

EINECSNumber : 211-207-2

TSCA : TSCA

SDS Documents