missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3-Trimethoxybenzene, 98+%

Manufacturer:  Alfa Aesar™ A1457236

 View more versions of this product

Catalog No. AAA1457236



Description

Description

Applications
1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Specifications

Specifications

1,2,3-Trimethoxybenzene
241°C
500g
C9H12O3
1910422
CRUILBNAQILVHZ-UHFFFAOYSA-N
1,2,3-trimethoxybenzene
12462
168.19
112°C (233°F)
43°C to 45°C
634-36-6
MFCD00008358
methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
COC1=C(C(=CC=C1)OC)OC
168.192
CHEBI:86529
≥98%

Chemical Identifiers

634-36-6
168.192
CRUILBNAQILVHZ-UHFFFAOYSA-N
12462
1,2,3-trimethoxybenzene
C9H12O3
MFCD00008358
methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
CHEBI:86529
COC1=C(C(=CC=C1)OC)OC
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents