Please login to your online account to display your discounted pricing

1,2,3-Trichloropropane, 98+%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  149555000

 View more versions of this product

Catalog No. AC149555000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 147.43
Physical Form Liquid
Quantity 500mL
Color Clear colorless to pale yellow
Beilstein 01, 106
Density 1.3860g/mL
CAS Min % 98.0
CAS Max % 100.0
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Linear Formula ClCH2CH(Cl)CH2Cl
Assay Percent Range 98+%
MDL Number MFCD00000946
Chemical Name or Material 1, 2, 3-Trichloropropane
CAS 96-18-4
Molecular Formula C3H5Cl3
Health Hazard 2 GHS H Statement:
May cause cancer.
May damage fertility.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1, 2, 3-Trichloropropane
CAS 96-18-4
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C3H5Cl3
Linear Formula ClCH2CH(Cl)CH2Cl
Formula Weight 147.43
MDL Number MFCD00000946
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
May damage fertility.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

SDS & Safety Documents