Please login to your online account to display your discounted pricing

1,2,3-Trichloropropane, 98+%, ACROS Organics™

$20.66 - $77.24

Chemical Identifiers

Chemical Name or Material 1, 2, 3-Trichloropropane
CAS 96-18-4
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C3H5Cl3
Linear Formula ClCH2CH(Cl)CH2Cl
Formula Weight 147.43
MDL Number MFCD00000946
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC149555000

  SDS
thermo fisher scientific
149555000
Glass bottle 500mL Each for $77.24

AC149551000

  SDS
thermo fisher scientific
AC149551000
Glass bottle 100mL Each for $27.35

AC149550050

  SDS
thermo fisher scientific
149550050
Glass bottle 5mL Each for $20.66
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Liquid
Formula Weight 147.43
Quantity 500mL
Color Clear colorless to pale yellow
Beilstein 01, 106
Density 1.3860g/mL
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula ClCH2CH(Cl)CH2Cl
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Chemical Name or Material 1, 2, 3-Trichloropropane
Molecular Formula C3H5Cl3
Infrared Spectrum Authentic
MDL Number MFCD00000946
CAS 96-18-4
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause cancer.
May damage fertility.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
May damage fertility.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.