Learn More
1,2,3-Trichloropropane, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L043120B
Description
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.
Solubility
Slightly soluble in water (2 g/L at 25°C).
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Specifications
1,2,3-Trichloropropane | |
-14°C | |
156°C to 158°C | |
C3H5Cl3 | |
MFCD00000946 | |
UN2810 | |
trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132 | |
CFXQEHVMCRXUSD-UHFFFAOYSA-N | |
1,2,3-trichloropropane | |
7285 | |
147.43 |
96-18-4 | |
1.385 | |
74°C (165°F) | |
1.4822 | |
1000 g | |
1732068 | |
Slightly soluble in water (2g/L at 25°C). | |
ClCC(Cl)CCl | |
147.42 | |
CHEBI:34036 | |
≥98% |
Chemical Identifiers
96-18-4 | |
147.42 | |
CFXQEHVMCRXUSD-UHFFFAOYSA-N | |
7285 | |
1,2,3-trichloropropane |
C3H5Cl3 | |
MFCD00000946 | |
trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132 | |
CHEBI:34036 | |
ClCC(Cl)CCl |
Safety and Handling
GHS H Statement
H350-H360-H302-H312-H332-H227
May cause cancer.
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
P201-P202-P210-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P370+P378q-P501c
H227-H302+H312-H350-H360F
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 202-486-1
RTECSNumber : TZ9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only