Please login to your online account to display your discounted pricing

1,2,3-Trichlorobenzene 99%, ACROS Organics™

$30.00 - $30.00

Chemical Identifiers

Chemical Name or Material 1, 2, 3-Trichlorobenzene
Name Note 99%
CAS 87-61-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3Cl3
Formula Weight 181.45
MDL Number MFCD00000537
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC139390050

  SDS
thermo fisher scientific
139390050
Glass bottle 5g Each for $30.00
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Solid
Formula Weight 181.45
Quantity 5g
Color White
EINECS Number 201-757-1
TSCA TSCA
Melting Point 50° to 54°C
RTECS Number DC2095000
Beilstein 05, 203
Merck Index 15, 9795
CAS Max % 100.0
Chemical Name or Material 1, 2, 3-Trichlorobenzene
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H3Cl3
Infrared Spectrum Authentic
MDL Number MFCD00000537
CAS 87-61-6
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.