missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3-Trichlorobenzene, 99%, Thermo Scientific Chemicals

$50.21 - $50.21

Chemical Identifiers

CAS 87-61-6
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000537
InChI Key RELMFMZEBKVZJC-UHFFFAOYSA-N
Synonym trichlorobenzene, vic-trichlorobenzene, 1,2,6-trichlorobenzene, benzene, 1,2,3-trichloro, benzene, trichloro, unii-nur9777ik4, ccris 5944, dsstox_cid_6193, dsstox_rid_78052, dsstox_gsid_26193
PubChem CID 6895
ChEBI CHEBI:35289
IUPAC Name 1,2,3-trichlorobenzene
SMILES ClC1=CC=CC(Cl)=C1Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2215422
View Documents Promotion Details
Thermo Scientific Chemicals
B2215422
100 g
Each for $50.21
 
Description

Description

Detoxification by catalytic hydrotreatment of 1,2,3-Trichlorobenzene is used for the disposal of hazardous organic waste liquids. As constituent of trichlorobenzene mixt used for termite control. As transformer fluid, dye carrier & solvent. Solvent for high melting products, Coolant in electrical installations and glass tempering. In polyester dyeing, lubricants, Heat transfer medium. As chemical intermediate for 2,3-dichlorophenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Detoxification by catalytic hydrotreatment of 1,2,3-Trichlorobenzene is used for the disposal of hazardous organic waste liquids. As constituent of trichlorobenzene mixt used for termite control. As transformer fluid, dye carrier & solvent. Solvent for high melting products, Coolant in electrical installations and glass tempering. In polyester dyeing, lubricants, Heat transfer medium. As chemical intermediate for 2,3-dichlorophenol.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

87-61-6
181.44
RELMFMZEBKVZJC-UHFFFAOYSA-N
6895
1,2,3-trichlorobenzene
C6H3Cl3
MFCD00000537
trichlorobenzene, vic-trichlorobenzene, 1,2,6-trichlorobenzene, benzene, 1,2,3-trichloro, benzene, trichloro, unii-nur9777ik4, ccris 5944, dsstox_cid_6193, dsstox_rid_78052, dsstox_gsid_26193
CHEBI:35289
ClC1=CC=CC(Cl)=C1Cl

Specifications

53°C to 55°C
1.69
100 g
MFCD00000537
956882
trichlorobenzene, vic-trichlorobenzene, 1,2,6-trichlorobenzene, benzene, 1,2,3-trichloro, benzene, trichloro, unii-nur9777ik4, ccris 5944, dsstox_cid_6193, dsstox_rid_78052, dsstox_gsid_26193
RELMFMZEBKVZJC-UHFFFAOYSA-N
1,2,3-trichlorobenzene
6895
181.45
1,2,3-Trichlorobenzene
87-61-6
218°C to 219°C
C6H3Cl3
UN3077
14,9630
Insoluble in water.
ClC1=CC=CC(Cl)=C1Cl
181.44
CHEBI:35289
99%
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 201-757-1

RTECSNumber : DC2095000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only