Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3-Trichloro-4-nitrobenzene, 97%

Chemical Identifiers

Chemical Name or Material 1,2,3-Trichloro-4-nitrobenzene
CAS 17700-09-3
Molecular Formula C6H2Cl3NO2
Formula Weight 226.45
MDL Number MFCD00007063
UN Number UN2811
Beilstein 2211951
Synonym 4-Nitro-1,2,3-trichlorobenze; 2,3,4-Trichloronitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1803809

 
alfa aesar™
A1803809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1803818

 
alfa aesar™
A1803818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.45
Chemical Name or Material 1,2,3-Trichloro-4-nitrobenzene
Assay 97%
Molecular Formula C6H2Cl3NO2
MDL Number MFCD00007063
CAS 17700-09-3
UN Number UN2811
Beilstein 2211951
Synonym 4-Nitro-1,2,3-trichlorobenze; 2,3,4-Trichloronitrobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 53° to 56°C
Flash Point >110°C (230°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number DA6612000
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.