missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3-Tribromopropane, 98%

$24.38 - $47.26

Chemical Identifiers

CAS 96-11-7
Molecular Formula C3H5Br3
Formula Weight 280.8
MDL Number MFCD00017884
Beilstein 1732082
Merck Index 14,9614
Chemical Name or Material 1,2,3-Tribromopropane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0375214 SDS Alfa Aesar™
L0375214
25g Each for $24.38
AAL0375222 SDS Alfa Aesar™
L0375222
100g Each for $47.26
Specifications Description & Specifications

Chemical Identifiers

96-11-7
280.8
1732082
1,2,3-Tribromopropane
C3H5Br3
MFCD00017884
14,9614

Specifications

96-11-7
280.8
1732082
16° to 17°C
25g
1,2,3-Tribromopropane
C3H5Br3
MFCD00017884
14,9614
220°C
93°C (199°F)
98%

Safety and Handling

GHS H Statement
H301-H341-H227-H315-H319
Toxic if swallowed.
Suspected of causing genetic defects.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P280-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : TZ8300000

TSCA : TSCA