Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3-Thiadiazole-4-carboxaldehyde, 98%

$343.26 - $343.26

Chemical Identifiers

Chemical Name or Material 1,2,3-Thiadiazole-4-carboxaldehyde
CAS 27643-15-8
Molecular Formula C3H2N2OS
Formula Weight 114.12
MDL Number MFCD02180762
Beilstein 113707
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2021103

 
alfa aesar™
L2021103
1g Each for $343.26
Description & Specifications

Specifications

Formula Weight 114.12
Chemical Name or Material 1,2,3-Thiadiazole-4-carboxaldehyde
Assay 98%
CAS 27643-15-8
Beilstein 113707
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C3H2N2OS
MDL Number MFCD02180762
Melting Point 84° to 86°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.