Please login to your online account to display your discounted pricing

1,2,3-Thiadiazole-4-carbaldehyde, Maybridge

Chemical Identifiers

Chemical Name or Material 1, 2, 3-Thiadiazole-4-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C3H2N2OS
Formula Weight 114.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP01373DA

  SDS
fisher scientific company
SP01373DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.13
Quantity 1g
Assay 97%
Chemical Name or Material 1, 2, 3-Thiadiazole-4-carbaldehyde
Molecular Formula C3H2N2OS
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 27643-15-8