Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3-Hexanetriol, 97%

$373.68 - $373.68

Chemical Identifiers

Chemical Name or Material 1,2,3-Hexanetriol
CAS 90325-47-6
Molecular Formula C6H14O3
Formula Weight 134.18
MDL Number MFCD00043071
Synonym 1,2,3-Trihydroxyhexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2718006

  SDS
alfa aesar™
H2718006
5g Each for $373.68
Description & Specifications

Specifications

Formula Weight 134.18
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
MDL Number MFCD00043071
Quantity 5g
Molecular Formula C6H14O3
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
CAS 90325-47-6
Health Hazard 1 Warning
Melting Point 64° to 67°C
Chemical Name or Material 1,2,3-Hexanetriol
Assay 97%
Synonym 1,2,3-Trihydroxyhexane
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.