Please login to your online account to display your discounted pricing

1-(2,3-Dimethylphenyl)piperazine, 97%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC136931000

 View more versions of this product

Catalog No. AC136931000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Formula Weight 190.29
Quantity 100g
Color Clear light yellow
Assay 98.5 to 101.5% Total base
Color Index No. 1-(2,3-Dimethylphenyl)piperazine
Density 1.0309g/mL
CAS Max % 100.0
Chemical Name or Material 1-(2, 3-Dimethylphenyl)piperazine
Refractive Index 1.5566 to 1.5606 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C12H18N2
CAS 1013-22-5
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-(2, 3-Dimethylphenyl)piperazine
CAS 1013-22-5
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5 to 101.5% Total base
Assay Percent Range 99%
Molecular Formula C12H18N2
Formula Weight 190.29
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

SDS & Safety Documents