Please login to your online account to display your discounted pricing

1-(2,3-Dimethylphenyl)piperazine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(2, 3-Dimethylphenyl)piperazine
CAS 1013-22-5
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5 to 101.5% Total base
Assay Percent Range 99%
Molecular Formula C12H18N2
Formula Weight 190.29
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC136931000

  SDS
thermo fisher scientific
AC136931000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC136930050

  SDS
fisher scientific acros
136930050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC136930250

  SDS
thermo fisher scientific
136930250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 2 GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Physical Form Viscous Liquid
Formula Weight 190.29
Quantity 100g
Color Clear light yellow
Assay 98.5 to 101.5% Total base
Color Index No. 1-(2,3-Dimethylphenyl)piperazine
Density 1.0309g/mL
CAS Max % 100.0
Refractive Index 1.5566 to 1.5606 (20°C, 589nm)
Chemical Name or Material 1-(2, 3-Dimethylphenyl)piperazine
Molecular Formula C12H18N2
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 1 Warning
CAS 1013-22-5
Assay Percent Range 99%
Safety and Handling

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.