Please login to your online account to display your discounted pricing

1-(2,3-Dichlorophenyl)ethan-1-one, 97%, Maybridge

$289.50 - $289.50

Chemical Identifiers

Chemical Name or Material 1-(2, 3-Dichlorophenyl)ethan-1-one
CAS 56041-57-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6Cl2O
Linear Formula Cl2 C6 H3 C (O) C H3
Formula Weight 189.04
MDL Number MFCD00052988
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00325EA

  SDS
fisher scientific company
TL00325EA
Amber Glass Bottle 10g Each for $289.50
Description & Specifications

Specifications

Formula Weight 189.04
Quantity 10g
Assay 97%
Density 1.293g/mL
CAS Min % 97.0
Chemical Name or Material 1-(2, 3-Dichlorophenyl)ethan-1-one
Linear Formula Cl2 C6 H3 C (O) C H3
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Molecular Formula C8H6Cl2O
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00052988
CAS 56041-57-7
Assay Percent Range 97%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.