Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3-Benzotriazin-4(3H)one, 97%

$40.38 - $129.43

Chemical Identifiers

Chemical Name or Material 1,2,3-Benzotriazin-4(3H)one
CAS 90-16-4
Molecular Formula C7H5N3O
Formula Weight 147.14
MDL Number MFCD00052387
Beilstein 124996
Synonym 4-Ketobenzo-1,2,3(3H)-triazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1979806

  SDS
alfa aesar™
A1979806
5g Each for $40.38

AAA1979814

  SDS
alfa aesar™
A1979814
25g Each for $129.43
Description & Specifications

Specifications

Formula Weight 147.14
Beilstein 124996
Synonym 4-Ketobenzo-1,2,3(3H)-triazine
Chemical Name or Material 1,2,3-Benzotriazin-4(3H)one
Molecular Formula C7H5N3O
TSCA TSCA
MDL Number MFCD00052387
Quantity 5g
Health Hazard 2 GHS H Statement
H301-H361-H332
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Harmful if inhaled.
Assay 97%
CAS 90-16-4
RTECS Number DM0800000
Melting Point 222° to 224°C
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P281-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
1,2,3-Benzotriazin-4(3H)one is used to prepare 3-methoxymethyl-3H-benzo[d][1,2,3]triazin-4-one by reacting with dimethoxymethane. Further, it undergoes thermal condensation with alfa-amino acids to give 3H-1,4-benzodiazepin-(1H,4H)-2,5-diones. In addition to this, it is employed as a reagent to synthesize 1,2,3-bBenzotriazin-4-one-arylpiperazine derivatives.

Solubility
Soluble in most organic solvents.

Notes
Incompatible with strong oxidizing agents, strong acid and bases.

Safety and Handling

Danger

GHS P Statement
P261-P281-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H361-H332
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Harmful if inhaled.