Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3-Benzenetricarboxylic acid hydrate, 99%

$224.51 - $224.51

Chemical Identifiers

Chemical Name or Material 1,2,3-Benzenetricarboxylic acid hydrate
CAS 732304-21-1
Molecular Formula C9H6O6·xH2O
Formula Weight 210.14 (Anhydrous)
MDL Number MFCD00149097
Beilstein 2214816
Synonym Hemimellitic acid hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1749114

  SDS
alfa aesar™
A1749114
25g Each for $224.51
Description & Specifications

Specifications

Formula Weight 210.14 (Anhydrous)
Chemical Name or Material 1,2,3-Benzenetricarboxylic acid hydrate
Assay 99%
Molecular Formula C9H6O6·xH2O
Melting Point ≈191°C (dec.)
MDL Number MFCD00149097
Quantity 25g
CAS 732304-21-1
Beilstein 2214816
Synonym Hemimellitic acid hydrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used in the synthesis and characterization and thermal behavior of hemimellitic acid complexes with lanthanides(III).

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.