missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose, 98%

Manufacturer:  Alfa Aesar™ J6693003

 View more versions of this product

Catalog No. AAJ6693003



Specifications

Specifications

1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose
White
1g
C34H28O9
1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-b-d-ribofuranose, 2s,3r,4r,5r-5-benzoyloxy methyl-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate, 2s,3r,4r,5r-2,4-bis benzoyloxy-5-benzoyloxy methyl-3-methyloxolan-3-yl benzoate, 1,2,3,5-tetra-o-benzoyl-2'-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d ribofuranose, 2-c-methyl-beta-d-ribofuranose 1,2,3,5-tetrabenzoate, 2-c-methyl-1,2,3,5-tetra-o-benzoyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofura
QJZSLTLDMBDKOU-VBHQRPIPSA-N
[(2R,3R,4R,5S)-3,4,5-tribenzoyloxy-4-methyloxolan-2-yl]methyl benzoate
10875560
Powder
+70° (c=1 in Chloroform)
147°C to 163°C
15397-15-6
MFCD07369658
Soluble in chloroform or acetonitrile
CC1(C(C(OC1OC(=O)C2=CC=CC=C2)COC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5
580.589
580.58
98%

Chemical Identifiers

15397-15-6
580.589
QJZSLTLDMBDKOU-VBHQRPIPSA-N
10875560
CC1(C(C(OC1OC(=O)C2=CC=CC=C2)COC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5
C34H28O9
MFCD07369658
1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-b-d-ribofuranose, 2s,3r,4r,5r-5-benzoyloxy methyl-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate, 2s,3r,4r,5r-2,4-bis benzoyloxy-5-benzoyloxy methyl-3-methyloxolan-3-yl benzoate, 1,2,3,5-tetra-o-benzoyl-2'-c-methyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d ribofuranose, 2-c-methyl-beta-d-ribofuranose 1,2,3,5-tetrabenzoate, 2-c-methyl-1,2,3,5-tetra-o-benzoyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-d-ribofuranose, 1,2,3,5-tetra-o-benzoyl-2-c-methyl-beta-d-ribofura
[(2R,3R,4R,5S)-3,4,5-tribenzoyloxy-4-methyloxolan-2-yl]methyl benzoate
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C

Documents