missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2:3,5-Di-O-isopropylidene-alpha-D-xylofuranose

$99.92 - $293.00

Chemical Identifiers

CAS 20881-04-3
Molecular Formula C11H18O5
Molecular Weight (g/mol) 230.26
MDL Number MFCD00063224
InChI Key NKZDPBSWYPINNF-BZNPZCIMSA-N
Synonym 1,2:3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1s,2r,6r,8r-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 6.4.0.0 2 ,? dodecane, xylofuranose, 1,2:3,5-di-o-isopropylidene-, .alpha.-d, .alpha.-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1,2,3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1-o,2-o:3-o,5-o-diisopropylidene-alpha-d-xylofuranose, .alpha.-d-xylofuranose, 1,2,3,5-bis-o-1-methylethylidene
PubChem CID 30337
IUPAC Name (3aR,4aR,8aS,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-3aH-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxine
SMILES CC1(OCC2C(O1)C3C(O2)OC(O3)(C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6650503 View Documents Alfa Aesar™
J6650503
1g Each for $99.92
 
AAJ6650506 View Documents Alfa Aesar™
J6650506
5g Each for $293.00
 
Specifications

Chemical Identifiers

20881-04-3
230.26
NKZDPBSWYPINNF-BZNPZCIMSA-N
30337
CC1(OCC2C(O1)C3C(O2)OC(O3)(C)C)C
C11H18O5
MFCD00063224
1,2:3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1s,2r,6r,8r-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 6.4.0.0 2 ,? dodecane, xylofuranose, 1,2:3,5-di-o-isopropylidene-, .alpha.-d, .alpha.-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1,2,3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1-o,2-o:3-o,5-o-diisopropylidene-alpha-d-xylofuranose, .alpha.-d-xylofuranose, 1,2,3,5-bis-o-1-methylethylidene
(3aR,4aR,8aS,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-3aH-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxine

Specifications

White
20881-04-3
MFCD00063224
NKZDPBSWYPINNF-BZNPZCIMSA-N
(3aR,4aR,8aS,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-3aH-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxine
30337
Powder
1g
C11H18O5
1,2:3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1s,2r,6r,8r-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 6.4.0.0 2 ,? dodecane, xylofuranose, 1,2:3,5-di-o-isopropylidene-, .alpha.-d, .alpha.-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1,2,3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1-o,2-o:3-o,5-o-diisopropylidene-alpha-d-xylofuranose, .alpha.-d-xylofuranose, 1,2,3,5-bis-o-1-methylethylidene
CC1(OCC2C(O1)C3C(O2)OC(O3)(C)C)C
230.26
230.26
1,2:3,5-Di-O-isopropylidene-alpha-D-xylofuranose