missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride, TCI America™

Manufacturer:  TCI America T272625G

 View more versions of this product

Catalog No. T272625G



Specifications

Specifications

1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride
228°C
13171-25-0
MFCD00243086
VYFLPFGUVGMBEP-UHFFFAOYSA-N
1-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride
83201
Crystal-Powder at 20°C
White
25g
C14H24Cl2N2O3
trimetazidine dihydrochloride, 1-2,3,4-trimethoxybenzyl piperazine dihydrochloride, kyurinett, trimetazidine hydrochloride, trimetajust, lubomail, vastarel, yosimilon, 1-2,3,4-trimethoxyphenyl methyl piperazine dihydrochloride, trimetazidine 2hcl
COC1=C(C(=C(C=C1)CN2CCNCC2)OC)OC.Cl.Cl
339.257
339.26
≥98.0% (HPLC,N)

Chemical Identifiers

13171-25-0
339.257
VYFLPFGUVGMBEP-UHFFFAOYSA-N
83201
COC1=C(C(=C(C=C1)CN2CCNCC2)OC)OC.Cl.Cl
C14H24Cl2N2O3
MFCD00243086
trimetazidine dihydrochloride, 1-2,3,4-trimethoxybenzyl piperazine dihydrochloride, kyurinett, trimetazidine hydrochloride, trimetajust, lubomail, vastarel, yosimilon, 1-2,3,4-trimethoxyphenyl methyl piperazine dihydrochloride, trimetazidine 2hcl
1-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : TM2976100

TSCA : No

Documents
Promotions

Promotions