Please login to your online account to display your discounted pricing

1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 85%, mixture of isomers, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC348070010

 View more versions of this product

Catalog No. AC348070010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥80% (GC)
Formula Weight 122.21
Physical Form Liquid
Quantity 1g
Color Light yellow
Density 0.808g/mL
CAS 858238
Chemical Name or Material 1,2,3,4-Tetramethyl-1,3-cyclopentadiene
Infrared Spectrum Authentic
Assay Percent Range 85%
Molecular Formula C9H14
Refractive Index 1.4700 to 1.4760 (20°C, 589nm)
Health Hazard 1 Warning

Specifications

Chemical Name or Material 1,2,3,4-Tetramethyl-1,3-cyclopentadiene
Assay ≥80% (GC)
Assay Percent Range 85%
Molecular Formula C9H14
Formula Weight 122.21
Accessories

Warning