missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3,4-Tetrahydroquinoxaline, 98%

Manufacturer:  Alfa Aesar™ H55473MD

 View more versions of this product

Catalog No. AAH55473MD



Specifications

Specifications

1,2,3,4-Tetrahydroquinoxaline
250mg
C8H10N2
1,2,3,4-terahydroquinoxaline
C1CNC2=CC=CC=C2N1
134.182
134.18
92°C to 98°C
3476-89-9
MFCD00047564
HORKYAIEVBUXGM-UHFFFAOYSA-N
1,2,3,4-tetrahydroquinoxaline
77028
98%

Chemical Identifiers

3476-89-9
134.182
HORKYAIEVBUXGM-UHFFFAOYSA-N
77028
C1CNC2=CC=CC=C2N1
C8H10N2
MFCD00047564
1,2,3,4-terahydroquinoxaline
1,2,3,4-tetrahydroquinoxaline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents