Learn More
1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 138275000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1,2,3,4-Tetrahydroquinoline | |
1.0600g/mL | |
100°C | |
97.5% min. (GC) | |
C9H11N | |
MFCD00006693 | |
20,262 | |
tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline | |
LBUJPTNKIBCYBY-UHFFFAOYSA-N | |
1,2,3,4-tetrahydroquinoline | |
69460 | |
CHEBI:213323 | |
98% |
635-46-1 | |
249.0°C | |
Authentic | |
Glass bottle | |
1.5930 to 1.5950 | |
500g | |
1.06 | |
Solubility in water: <1g/L (20°C). Other solubilities: soluble in ethanol | |
C1CNC2=CC=CC=C2C1 | |
133.19 | |
12.5 mPa.s (20°C) | |
133.19 |
Safety and Handling
GHS H Statement
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 211-237-6
Recommended Storage : Product may darken during storage
RUO â Research Use Only