missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydronaphthalene, 98+%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 146730010

 View more versions of this product

Catalog No. AC146730010



Specifications

Specifications

1,2,3,4-Tetrahydronaphthalene
77°C
1.5400 to 1.5420
2.2 mPa.s (20°C)
-35.0°C
119-64-2
100.0
C10H12
MFCD00001733
01,1142
bacticin, benzocyclohexane, naphthalene 1,2,3,4-tetrahydride, naphthalene, 1,2,3,4-tetrahydro, tetrahydronaphthalene, tetralene, tetralin, tetralina, tetraline, tetranap
CXWXQJXEFPUFDZ-UHFFFAOYSA-N
1,2,3,4-tetrahydronaphthalene
8404
132.2
Pure
0.9730g/mL
Glass bottle
0.973
207.0°C
1L
98.0
98% min. (GC)
C10H12
05, IV, 1388
15, 9368
Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
C1CCC2=CC=CC=C2C1
132.2
CHEBI:35008
98+%

Chemical Identifiers

119-64-2
132.2
CXWXQJXEFPUFDZ-UHFFFAOYSA-N
8404
1,2,3,4-tetrahydronaphthalene
C10H12
MFCD00001733
bacticin, benzocyclohexane, naphthalene 1,2,3,4-tetrahydride, naphthalene, 1,2,3,4-tetrahydro, tetrahydronaphthalene, tetralene, tetralin, tetralina, tetraline, tetranap
CHEBI:35008
C1CCC2=CC=CC=C2C1
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May form explosive peroxides.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Warning

EINECSNumber : 204-340-2

SDS Documents