missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydronaphthalene-1-carboxylic Acid, TCI America™

Manufacturer:  TCI America T290325G

 View more versions of this product

Catalog No. T290325G



Specifications

Specifications

1,2,3,4-Tetrahydronaphthalene-1-carboxylic Acid
Undesignated
25g
C11H12O2
1,2,3,4-tetrahydro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydro, tetralin-1-carboxylic acid, 1,2,3,4-tetrahedro-naphthoic acid, 1,2,3,4-tetrahydro-naphthalene-1-carboxylic acid, chembl81606, 1-tetralincarboxylic acid, 1,2,3,4-tetrahydro-1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydronaphthalenecarboxylic acid, tetrahydronaphthoic acid
C1CC(C2=CC=CC=C2C1)C(=O)O
176.215
176.22
≥98.0% (GC,T)
135°C
84°C
1914-65-4
MFCD02179127
VDLWTJCSPSUGOA-UHFFFAOYSA-N
1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
12598146
Crystal-Powder at 20°C

Chemical Identifiers

1914-65-4
176.215
VDLWTJCSPSUGOA-UHFFFAOYSA-N
12598146
C1CC(C2=CC=CC=C2C1)C(=O)O
C11H12O2
MFCD02179127
1,2,3,4-tetrahydro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydro, tetralin-1-carboxylic acid, 1,2,3,4-tetrahedro-naphthoic acid, 1,2,3,4-tetrahydro-naphthalene-1-carboxylic acid, chembl81606, 1-tetralincarboxylic acid, 1,2,3,4-tetrahydro-1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydronaphthalenecarboxylic acid, tetrahydronaphthoic acid
1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1181

TSCA : No

Documents