missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydroisoquinoline, TCI America™

Manufacturer:  TCI America T1676100G

 View more versions of this product

Catalog No. T1676100G



Specifications

Specifications

1,2,3,4-Tetrahydroisoquinoline
Yellow
100g
C9H11N
tetrahydroisoquinoline, 1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro isoquinoline, 1,2,3,4-tetrahydro-2-isoquinoline, 1,2,3,4-tetrahydro-2-azanaphthalene, unii-56w89fbx3e, chembl14346, 3,4-dihydro-1h-isoquinoline, tetrahydroisoquinolin
C1CNCC2=CC=CC=C21
133.194
133.19
≥95.0% (GC)
233°C
-30°C
91-21-4
MFCD00006896
UWYZHKAOTLEWKK-UHFFFAOYSA-N
1,2,3,4-tetrahydroisoquinoline
7046
Clear Liquid at 20°C

Chemical Identifiers

91-21-4
133.194
UWYZHKAOTLEWKK-UHFFFAOYSA-N
7046
C1CNCC2=CC=CC=C21
C9H11N
MFCD00006896
tetrahydroisoquinoline, 1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro isoquinoline, 1,2,3,4-tetrahydro-2-isoquinoline, 1,2,3,4-tetrahydro-2-azanaphthalene, unii-56w89fbx3e, chembl14346, 3,4-dihydro-1h-isoquinoline, tetrahydroisoquinolin
1,2,3,4-tetrahydroisoquinoline
Safety and Handling

Safety and Handling

RTECSNumber : NX4900000

TSCA : Yes

Documents