Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydroisoquinoline hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline hydrochloride
CAS 14099-81-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H11N·ClH
Formula Weight 169.65
MDL Number MFCD00012740
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC370000250

  SDS
thermo fisher scientific
AC370000250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 169.65
Physical Form Crystalline Powder
Quantity 25g
Color White to off-white
Melting Point 198° to 202°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Packaging Glass bottle
MDL Number MFCD00012740
CAS Min % 97.5
Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline hydrochloride
Assay Percent Range 98%
CAS 14099-81-1
Molecular Formula C9H11N·ClH
Infrared Spectrum Authentic