missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydroisoquinoline, 95%, ACROS Organics™

Manufacturer:  Acros Organics 138201000

 View more versions of this product

Catalog No. AC138201000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Yellow to Brown
1,2,3,4-Tetrahydroisoquinoline
Authentic
1.5665 to 1.5685 (20°C, 589nm)
94.0
95%
MFCD00006896
1 pound not2 pound not3 pound not4-tetrahydro-isoquinoline, 5-20-06-00320 beilstein handbook reference, 1234-tetrahydro-2-azanaphthalene, 1 2 3 4-tetrahydro-isoquinoline, 1 2 3 4-tetrahydroisoquinoline, isoquinoline 1234-tetrahydro, 1234-tetrahydro-2-isoquinoline, 1 2 3 4 tetrahydroisoquinoline, 1234-tetrahydro-isoquinoline
C1CNCC2=CC=CC=C21
133.194
133.19
≥94%
100g
1.06g/mL
Glass bottle
91-21-4
100.0
C9H11N
20, 275
UWYZHKAOTLEWKK-UHFFFAOYSA-N
1,2,3,4-tetrahydroisoquinoline
7046
Liquid

Chemical Identifiers

91-21-4
133.194
UWYZHKAOTLEWKK-UHFFFAOYSA-N
7046
C1CNCC2=CC=CC=C21
C9H11N
MFCD00006896
1 pound not2 pound not3 pound not4-tetrahydro-isoquinoline, 5-20-06-00320 beilstein handbook reference, 1234-tetrahydro-2-azanaphthalene, 1 2 3 4-tetrahydro-isoquinoline, 1 2 3 4-tetrahydroisoquinoline, isoquinoline 1234-tetrahydro, 1234-tetrahydro-2-isoquinoline, 1 2 3 4 tetrahydroisoquinoline, 1234-tetrahydro-isoquinoline
1,2,3,4-tetrahydroisoquinoline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents