Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1,2,3,4-Tetrahydroisoquinoline-3-methanol, 98%

$369.16 - $369.16

Chemical Identifiers

Chemical Name or Material (+/-)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
CAS 63006-93-9
Molecular Formula C10H13NO
Formula Weight 163.22
MDL Number MFCD02259418
Beilstein 140470
Synonym (+/-)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6593303

  SDS
alfa aesar™
H6593303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6593306

  SDS
alfa aesar™
H6593306
5g Each for $369.16
Description & Specifications

Specifications

Formula Weight 163.22
Physical Form Crystalline
Chemical Name or Material (+/-)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
CAS 63006-93-9
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H13NO
MDL Number MFCD02259418
Quantity 1g
Assay 98%
Beilstein 140470
Health Hazard 1 Warning
Synonym (+/-)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. It is also inhibitors of phenylethanolamine N-methyltransferase.

Solubility
Soluble in dichloromethane. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.