Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydrocarbazole, 99%, Acros Organics


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  138171000

 View more versions of this product

Catalog No. AC138171000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 171.24
Quantity 100g
Color Beige
TSCA TSCA
Melting Point 118° to 120°C
EINECS Number 213-385-7
RTECS Number FE6300000
Beilstein 20, 416
CAS Max % 100.0
Chemical Name or Material 1,2,3,4-Tetrahydrocarbazole
Packaging Glass bottle
Assay Percent Range 99%
Infrared Spectrum Authentic
Name Note 99%
Molecular Formula C12H13N
MDL Number MFCD00004959
CAS 942-01-8
CAS Min % 98.5

Specifications

Chemical Name or Material 1,2,3,4-Tetrahydrocarbazole
Name Note 99%
CAS 942-01-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H13N
Formula Weight 171.24
MDL Number MFCD00004959