Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3,4-Tetrahydrocarbazole, 99%

$58.15 - $155.20

Chemical Identifiers

Chemical Name or Material 1,2,3,4-Tetrahydrocarbazole
CAS 942-01-8
Molecular Formula C12H13N
Formula Weight 171.24
MDL Number MFCD00004959
Beilstein 133771
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2365714

  SDS
alfa aesar™
B2365714
25g Each for $58.15

AAB2365722

  SDS
alfa aesar™
B2365722
100g Each for $155.20
Description & Specifications

Specifications

Formula Weight 171.24
Chemical Name or Material 1,2,3,4-Tetrahydrocarbazole
Assay 99%
Molecular Formula C12H13N
MDL Number MFCD00004959
CAS 942-01-8
Beilstein 133771
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 325° to 330°C
Melting Point 118° to 120°C
Quantity 25g
RTECS Number FE6300000

Applications
It finds its application as a pharmaceutical intermediate and in the synthesis of aspidospermidine alkaloids.

Solubility
Insoluble in water. Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.