missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydro-9-methylcarbazol-4-one, TCI America™

Manufacturer:  TCI America T294225G

 View more versions of this product

Catalog No. T294225G



Specifications

Specifications

1,2,3,4-Tetrahydro-9-methylcarbazol-4-one
201°C
27387-31-1
MFCD00173748
HHJUJCWZKJMCLC-UHFFFAOYSA-N
9-methyl-2,3-dihydro-1H-carbazol-4-one
598875
Crystal-Powder at 20°C
White-Yellow
25g
C13H13NO
1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole
CN1C2=C(C(=O)CCC2)C3=CC=CC=C31
199.253
199.25
≥98.0% (GC)

Chemical Identifiers

27387-31-1
199.253
HHJUJCWZKJMCLC-UHFFFAOYSA-N
598875
CN1C2=C(C(=O)CCC2)C3=CC=CC=C31
C13H13NO
MFCD00173748
1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole
9-methyl-2,3-dihydro-1H-carbazol-4-one
Safety and Handling

Safety and Handling

TSCA : No

Documents