missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydro-2-naphthol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC271880050

 View more versions of this product

Catalog No. AC271880050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1,2,3,4-Tetrahydro-2-naphthol
Glass bottle
140°C (12.0mmHg)
15.5°C
530-91-6
100.0
C10H12O
15, 9369
JWQYZECMEPOAPF-UHFFFAOYSA-N
1,2,3,4-tetrahydronaphthalen-2-ol
10747
Viscous Liquid
Authentic
1.5620 to 1.564
Brown to Yellow
5g
96.0
96% min. (GC)
MFCD00045575
.beta.-tetralol, 1,2,3,4-tetrahydro-2-naphthol, 2-hydroxytetralin, 2-hydroxytetraline, 2-naphthalenol, 1,2,3,4-tetrahydro, 2-naphthol, 1,2,3,4-tetrahydro, 2-tetralinol, ac-tetrahydro-beta-naphthol, beta-tetralol, tetrahydronaphthol-2
C1CC2=CC=CC=C2CC1O
148.2
148.2
97%

Chemical Identifiers

530-91-6
148.2
JWQYZECMEPOAPF-UHFFFAOYSA-N
10747
C1CC2=CC=CC=C2CC1O
C10H12O
MFCD00045575
.beta.-tetralol, 1,2,3,4-tetrahydro-2-naphthol, 2-hydroxytetralin, 2-hydroxytetraline, 2-naphthalenol, 1,2,3,4-tetrahydro, 2-naphthol, 1,2,3,4-tetrahydro, 2-tetralinol, ac-tetrahydro-beta-naphthol, beta-tetralol, tetrahydronaphthol-2
1,2,3,4-tetrahydronaphthalen-2-ol
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-497-8

SDS Documents