missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrafluorobenzene, TCI America™

Manufacturer:  TCI America T241025G

 View more versions of this product

Catalog No. T241025G



Specifications

Specifications

1,2,3,4-Tetrafluorobenzene
Yellow
25g
C6H2F4
1993
SOZFIIXUNAKEJP-UHFFFAOYSA-N
1,2,3,4-tetrafluorobenzene
11084
Clear Liquid at 20°C
94°C
-42°C
551-62-2
MFCD00000285
tetrafluorobenzene, 1,2,3,4-tetrafluoro-benzene, unii-66365s2rfq, benzene, 1,2,3,4-tetrafluoro, 2,3,4,5-tetrafluorobenzene, benzene, tetrafluoro, pubchem1061, acmc-1ao1z, 1,3,4-tetrafluorobenzene
C1=CC(=C(C(=C1F)F)F)F
150.076
150.08
≥98.0% (GC)

Chemical Identifiers

551-62-2
150.076
SOZFIIXUNAKEJP-UHFFFAOYSA-N
11084
C1=CC(=C(C(=C1F)F)F)F
C6H2F4
MFCD00000285
tetrafluorobenzene, 1,2,3,4-tetrafluoro-benzene, unii-66365s2rfq, benzene, 1,2,3,4-tetrafluoro, 2,3,4,5-tetrafluorobenzene, benzene, tetrafluoro, pubchem1061, acmc-1ao1z, 1,3,4-tetrafluorobenzene
1,2,3,4-tetrafluorobenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents