missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3,4-Tetra-O-acetyl-beta-D-ribopyranose, 98%

$39.41 - $157.59

Chemical Identifiers

CAS 4049-34-7
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00006596
InChI Key MJOQJPYNENPSSS-LPWJVIDDSA-N
Synonym 1,2,3,4-tetra-o-acetyl-, 1,2,3,4-tetra-o-acetyl-b-d-ribopyranose, 2s,3r,4r,5r-2,4,5-tris acetyloxy oxan-3-yl acetate, 2s,3r,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 3r,4r,5r,6s-4,5,6-triacetyloxyoxan-3-yl acetate, a-d-ribopyranose, b-d-ribopyranose 1,2,3,4-tetraacetate, beta-d-ribopyranose 1,2,3,4-tetraacetate, beta-d-ribopyranose tetraacetate, mjoqjpynenpsss-lpwjviddsa, tetra-o-acetyl-beta-d-ribopyranose
PubChem CID 97336
IUPAC Name [(3R,4R,5R,6S)-4,5,6-triacetyloxyoxan-3-yl] acetate
SMILES CC(=O)OC1COC(C(C1OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66623MD
View Documents
Alfa Aesar™
J66623MD
250mg Each for $39.41
 
AAJ6662303
View Documents
Alfa Aesar™
J6662303
1g Each for $157.59
 
Specifications

Chemical Identifiers

4049-34-7
318.278
MJOQJPYNENPSSS-LPWJVIDDSA-N
97336
CC(=O)OC1COC(C(C1OC(=O)C)OC(=O)C)OC(=O)C
C13H18O9
MFCD00006596
1,2,3,4-tetra-o-acetyl-, 1,2,3,4-tetra-o-acetyl-b-d-ribopyranose, 2s,3r,4r,5r-2,4,5-tris acetyloxy oxan-3-yl acetate, 2s,3r,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 3r,4r,5r,6s-4,5,6-triacetyloxyoxan-3-yl acetate, a-d-ribopyranose, b-d-ribopyranose 1,2,3,4-tetraacetate, beta-d-ribopyranose 1,2,3,4-tetraacetate, beta-d-ribopyranose tetraacetate, mjoqjpynenpsss-lpwjviddsa, tetra-o-acetyl-beta-d-ribopyranose
[(3R,4R,5R,6S)-4,5,6-triacetyloxyoxan-3-yl] acetate

Specifications

+55° (c=3 in Methanol)
110°C to 112°C
4049-34-7
MFCD00006596
MJOQJPYNENPSSS-LPWJVIDDSA-N
[(3R,4R,5R,6S)-4,5,6-triacetyloxyoxan-3-yl] acetate
97336
Powder
1,2,3,4-Tetra-O-acetyl-beta-D-ribopyranose
White
250mg
C13H18O9
1,2,3,4-tetra-o-acetyl-, 1,2,3,4-tetra-o-acetyl-b-d-ribopyranose, 2s,3r,4r,5r-2,4,5-tris acetyloxy oxan-3-yl acetate, 2s,3r,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 3r,4r,5r,6s-4,5,6-triacetyloxyoxan-3-yl acetate, a-d-ribopyranose, b-d-ribopyranose 1,2,3,4-tetraacetate, beta-d-ribopyranose 1,2,3,4-tetraacetate, beta-d-ribopyranose tetraacetate, mjoqjpynenpsss-lpwjviddsa, tetra-o-acetyl-beta-d-ribopyranose
CC(=O)OC1COC(C(C1OC(=O)C)OC(=O)C)OC(=O)C
318.278
318.28
98%
Safety and Handling

Safety and Handling

EINECSNumber : 223-750-2

TSCA : No

Recommended Storage : Ambient temperatures