Learn More
1,2,3,4-Butanetetracarboxylic acid, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 366761000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2,3,4-Butanetetracarboxylic acid | |
1703-58-8 | |
100.0 | |
White | |
99+% | |
C8H10O8 | |
100 g | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O | |
234.16 | |
234.16 | |
Crystalline Powder |
98% | |
97.5 | |
195.0°C to 197.0°C | |
Authentic | |
Glass bottle | |
MFCD00002722 | |
1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa | |
GGAUUQHSCNMCAU-UHFFFAOYSA-N | |
butane-1,2,3,4-tetracarboxylic acid | |
15560 | |
99+% |
Chemical Identifiers
1703-58-8 | |
234.16 | |
GGAUUQHSCNMCAU-UHFFFAOYSA-N | |
15560 | |
C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O |
C8H10O8 | |
MFCD00002722 | |
1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa | |
butane-1,2,3,4-tetracarboxylic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 216-938-
RUO – Research Use Only