Learn More
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, CTC-Q-PHOS, 95%, Thermo Scientific Chemicals
Heck and Suzuki reactions
Supplier: Thermo Scientific Chemicals 04461802
Description
Heck and Suzuki reactions1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene is a sterically hindered phosphine ligand, which is used in palladium-catalyzed amination, arylation and etherification reactions of aryl chlorides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
1,2,3,4,5-Pentaphenyl-1'- (di-tert-butylphosphino)ferrocene | |
312959-24-3 | |
MFCD03788940 | |
Soluble in ether,benzene,toluene and terahydrofuran. | |
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene | |
710.72 | |
Powder |
CTC-Q-PHOS | |
C48H47FeP | |
0.5 g | |
[Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 | |
710.72 | |
95% |
Chemical Identifiers
312959-24-3 | |
710.72 | |
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene |
C48H47FeP | |
MFCD03788940 | |
[Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only