Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, CTC-Q-PHOS, 95%

Heck and Suzuki reactions

$111.91 - $394.05

Chemical Identifiers

Chemical Name or Material 1,2,3,4,5-Pentaphenyl-1'- (di-tert-butylphosphino)ferrocene
CAS 312959-24-3
Name Note CTC-Q-PHOS
Molecular Formula C48H47FeP
Formula Weight 710.72
MDL Number MFCD03788940
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4461877

 
alfa aesar™
4461877
0.1g Each for $111.91

AA4461802

 
alfa aesar™
4461802
0.5g Each for $394.05
Description & Specifications

Specifications

Formula Weight 710.72
Name Note CTC-Q-PHOS
Chemical Name or Material 1,2,3,4,5-Pentaphenyl-1'- (di-tert-butylphosphino)ferrocene
Physical Form Powder
Assay 95%
Molecular Formula C48H47FeP
Quantity 0.1g
MDL Number MFCD03788940
CAS 312959-24-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under Argon

  • Packaged under Argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Packaged under Argon