missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,3,4,5,6-Hexabromocyclohexane

Potently and directly inhibits JAK2 tyrosine kinase autophosphorylation

Manufacturer:  Alfa Aesar™ J6085306

 View more versions of this product

Catalog No. AAJ6085306



Specifications

Specifications

1,2,3,4,5,6-Hexabromocyclohexane
5g
C6H6Br6
benzene hexabromide, cyclohexane, 1,2,3,4,5,6-hexabromo, jak2 inhibitor ii, acmc-1bqjt, trans-alpha-benzene hexabromide, cyclohexane,2,3,4,5,6-hexabromo, 1,2,3,4,5,6-hexabromo-cyclohexane, #, 1,2,3,4,5,6-hexakis bromanyl cyclohexane, 1,2,3,4,5,6-hexabromocyclohexane hplc
C1(C(C(C(C(C1Br)Br)Br)Br)Br)Br
557.538
557.5
White
1837-91-8
MFCD00059127
QFQZKISCBJKVHI-UHFFFAOYSA-N
1,2,3,4,5,6-hexabromocyclohexane
74603
Solid

Chemical Identifiers

1837-91-8
557.538
QFQZKISCBJKVHI-UHFFFAOYSA-N
74603
C1(C(C(C(C(C1Br)Br)Br)Br)Br)Br
C6H6Br6
MFCD00059127
benzene hexabromide, cyclohexane, 1,2,3,4,5,6-hexabromo, jak2 inhibitor ii, acmc-1bqjt, trans-alpha-benzene hexabromide, cyclohexane,2,3,4,5,6-hexabromo, 1,2,3,4,5,6-hexabromo-cyclohexane, #, 1,2,3,4,5,6-hexakis bromanyl cyclohexane, 1,2,3,4,5,6-hexabromocyclohexane hplc
1,2,3,4,5,6-hexabromocyclohexane
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents