missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[2-(2-Hydroxyethoxy)ethyl]piperazine, 95%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5503403

 View more versions of this product

Catalog No. AAH5503403



Specifications

Specifications

1-[2-(2-Hydroxyethoxy)ethyl]piperazine
112°C to 114°C (0.25mmHg)
13349-82-1
MFCD00074931
FLNQAPQQAZVRDA-UHFFFAOYSA-N
2-(2-piperazin-1-ylethoxy)ethanol
139436
95%
113°C (235°F)
1g
C8H18N2O2
1-2-2-hydroxyethoxy ethyl piperazine
C1CN(CCN1)CCOCCO
174.244
174.24

Chemical Identifiers

13349-82-1
174.244
FLNQAPQQAZVRDA-UHFFFAOYSA-N
139436
C1CN(CCN1)CCOCCO
C8H18N2O2
MFCD00074931
1-2-2-hydroxyethoxy ethyl piperazine
2-(2-piperazin-1-ylethoxy)ethanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents