Please login to your online account to display your discounted pricing

1-[2-(2,5-Dimethyl-1H-pyrrol-1-yl)ethyl]piperazine, ≥90%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00470DA

 View more versions of this product

Catalog No. MO00470DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 207.32
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
MDL Number MFCD03407319
CAS 5059-36-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-[2-(2, 5-Dimethyl-1H-pyrrol-1-yl)ethyl]piperazine
Assay Percent Range 97%
Molecular Formula C12H21N3

Specifications

Chemical Name or Material 1-[2-(2, 5-Dimethyl-1H-pyrrol-1-yl)ethyl]piperazine
CAS 5059-36-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H21N3
Formula Weight 207.32
MDL Number MFCD03407319
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents