Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulfonate, 98%

Chemical Identifiers

Chemical Name or Material 1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulfonate
CAS 86386-77-8
Formula Weight 333.31
MDL Number MFCD11111978
Synonym 1-[2-(2,4-difluorophenyl)-2-oxiranylmethyl]-1H-1,2,4-triazole methanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6178203

 
alfa aesar™
H6178203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6178206

 
alfa aesar™
H6178206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 333.31
Chemical Name or Material 1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulfonate
Physical Form Solid
Assay 98%
MDL Number MFCD11111978
CAS 86386-77-8
Synonym 1-[2-(2,4-difluorophenyl)-2-oxiranylmethyl]-1H-1,2,4-triazole methanesulfonate
Quantity 1g